This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343880_g1
See other CD200R1 GE Assays ›
Gene Symbol:
Cd200r1
Gene Name:
CD200 receptor 1
Gene Aliases:
CD200R, Mox2r, OX2R
Chromosome Location:
Chr.16: 44765736 - 44794977 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343880_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd200r1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021325.3 NP_067300.1 5 - 6 1064 162
GenBank mRNA AK089173.1 - 5 - 6 1001 162
AK154325.1 - 5 - 6 1055 162
AK154722.1 - 5 - 6 1023 162
AK171130.1 - 5 - 6 996 162
AK171590.1 - 5 - 6 1000 162
BC052682.1 - 5 - 6 1022 30072503 162

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000662.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd200r1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Process(es)

receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005813.mm.1 NM_021325.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005813.mm.1 NM_021325.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600005813.mm.1 TC1600000662.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline