This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343889_g1
See other CD200R3 GE Assays ›
Gene Symbol:
Cd200r3
Gene Name:
CD200 receptor 3
Gene Aliases:
4733401I18Rik, 4833409J19Rik, AI505817, BB106877, mCD200RLb
Chromosome Location:
Chr.16: 44943596 - 45058365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343889_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd200r3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001128132.1 NP_001121604.1 3 - 4 584 70
NM_029018.4 NP_083294.2 3 - 4 584 70
GenBank mRNA AK170363.1 - 3 - 4 514 70
AY230199.1 - 3 - 4 558 70
AY703837.1 - 3 - 4 489 70
AY703838.1 - 3 - 4 489 70
AY703839.1 - 3 - 4 489 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000666.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd200r3

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Process(es)

receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005847.mm.1 NM_001128132.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005847.mm.1 NM_001128132.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005847.mm.1 NM_029018.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005847.mm.1 NM_029018.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005847.mm.1 TC1600000666.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline