This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343890_m1
See other CD200R3 GE Assays ›
Gene Symbol:
Cd200r3
Gene Name:
CD200 receptor 3
Gene Aliases:
4733401I18Rik, 4833409J19Rik, AI505817, BB106877, mCD200RLb
Chromosome Location:
Chr.16: 44943596 - 45058365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343890_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd200r3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001128132.1 NP_001121604.1 4 - 5 809 95
NM_029018.4 NP_083294.2 4 - 5 809 95
GenBank mRNA AK170363.1 - 4 - 5 740 95
AY230199.1 - 4 - 5 783 95
AY703837.1 - 4 - 5 714 95
AY703838.1 - 4 - 5 714 95
AY703839.1 - 4 - 5 714 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000666.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd200r3

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Process(es)

receptor activity
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005857.mm.1 NM_027578.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005857.mm.1 NM_027578.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005857.mm.1 NM_029018.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005857.mm.1 NM_029018.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005857.mm.1 TC1600000666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005858.mm.1 NM_027578.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005858.mm.1 NM_027578.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005858.mm.1 NM_029018.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005858.mm.1 NM_029018.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005858.mm.1 TC1600000666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005855.mm.1 NM_001128132.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005855.mm.1 NM_001128132.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005855.mm.1 NM_001128133.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005855.mm.1 NM_001128133.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005855.mm.1 TC1600000666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 NM_001128132.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 NM_001128132.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 NM_001128133.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 NM_001128133.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 NM_027578.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 NM_027578.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 NM_029018.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 NM_029018.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005848.mm.1 TC1600000666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005854.mm.1 NM_001128132.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005854.mm.1 NM_001128132.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005854.mm.1 NM_001128133.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005854.mm.1 NM_001128133.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005854.mm.1 TC1600000666.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 363000 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline