Assay Details
| Gene Symbol: Adprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007414.3 | NP_031440.1 | 3 - 4 | 879 | 74 | |
| GenBank mRNA | AF244347.1 | - | 848 | 74 | ||
| AK010501.1 | - | 3 - 4 | 844 | 74 | ||
| AK146280.1 | - | 3 - 4 | 849 | 74 | ||
| AK153498.1 | - | 3 - 4 | 844 | 74 | ||
| AK171798.1 | - | 3 - 4 | 849 | 74 | ||
| AK178645.1 | - | 1 - 2 | 137 | 74 | ||
| BC003437.1 | - | 3 - 4 | 837 | 2647247 | 74 | |
| BC058232.1 | - | 3 - 4 | 836 | 74 | ||
| L13290.1 | - | 2 - 3 | 676 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686071 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686023 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686022 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686068 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686020 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686021 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686065 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686070 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686019 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686069 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686066 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504478 | AK010501.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504478 | AK010501.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881748 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110366 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163650 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504478 | AK010501.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504478 | AK010501.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686100 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163650 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08569196 | - | - |
| Whole Mouse Genome G4122A | A_52_P504478 | AK010501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P504478 | AK010501.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P289022 | AF244347.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338615 | BC058232.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338615 | L13290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338615 | AK146280.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338615 | AK153498.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338615 | AK171798.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106602 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12638896 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12711588 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12711580 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12711570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12711578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12638884 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12638889 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12711583 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686020 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686021 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686019 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686070 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686065 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686071 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686068 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686023 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686069 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30685981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30685996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30685994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30685991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30685986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12843716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12600312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12766135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12534814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12819575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12864547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12612179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12760865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12628875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12600315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686026 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10439457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439461 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439462 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439460 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448333_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460183_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10439455 | - | - |
| Mouse Genome 430 2.0 Array | 1448333_at | - | - |
| Mouse Genome 430 2.0 Array | 1448333_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460183_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460183_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11544_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11544_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600013125.mm.1 | NM_007414.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013125.mm.1 | NM_007414.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013125.mm.1 | TC1600001605.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448333_at | - | - |
| Mouse Expression Set 430 | 1448333_PM_at | - | - |
| Mouse Expression Set 430 | 1460183_PM_at | - | - |
| Mouse Expression Set 430 | 1460183_at | - | - |
| Murine Genome U74v2 Set | 93540_PM_at | - | - |
| Murine Genome U74v2 Set | 93540_at | - | - |
| Murine Genome U74v2 Set | 169712_PM_at | - | - |
| Murine Genome U74v2 Set | 169712_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448333_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460183_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460183_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448333_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460183_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460183_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448333_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448333_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3840035 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4780044 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4780044 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3840035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003840035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780044 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684684 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2595704 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684684 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2595704 | - | - |
| mouse-6 v1.1 (Illumina) | scl011544.1_20-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0011544.2_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780044 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3840035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl011544.1_20-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0011544.2_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780044 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2595704 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2595704 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl011544.1_20-S | - | - |

Adprh
Genomic Map