This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01344325_g1
See other POLR2H GE Assays ›
Gene Symbol:
Polr2h
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide H
Gene Aliases:
-
Chromosome Location:
Chr.16: 20717826 - 20722272 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01344325_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145632.2 NP_663607.1 1 - 2 169 118
GenBank mRNA AK146429.1 - 1 - 2 194 118
AK161617.1 - 1 - 2 190 118
BC002306.1 - 1 - 2 169 3591968 118
BC092087.1 - 1 - 2 79 5042770 118
BC125612.1 - 1 - 2 153 40130127 118
BC125638.1 - 1 - 2 153 40130153 118

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr2h

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase II promoter
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0