This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01344328_g1
See other POLR2H GE Assays ›
Gene Symbol:
Polr2h
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide H
Gene Aliases:
-
Chromosome Location:
Chr.16: 20717826 - 20722272 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01344328_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145632.2 NP_663607.1 4 - 5 432 82
GenBank mRNA AK146429.1 - 4 - 5 457 82
AK161617.1 - 4 - 5 453 82
BC092087.1 - 4 - 5 342 5042770 82
BC125612.1 - 4 - 5 416 40130127 82
BC125638.1 - 4 - 5 416 40130153 82

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr2h

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase II promoter
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589176 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589177 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589175 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966337 AK146429.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966337 AK161617.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966337 NM_145632.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966337 BC092087.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502456 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699365 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387876 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122201 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966337 AK146429.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966337 AK161617.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966337 NM_145632.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966337 BC092087.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108731 BC002306.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589181 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589193 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589180 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589178 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589186 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589184 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589189 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589183 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589185 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589177 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589195 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589192 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30589175 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03144671 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122201 - -
Whole Mouse Genome G4122A A_51_P108731 BC002306.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108041 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122201 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12672584 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589177 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589175 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30589176 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589150 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589180 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589192 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589109 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589130 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589178 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589110 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589115 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589202 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589152 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589244 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589184 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589206 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589156 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589108 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589173 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589125 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589160 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589256 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589172 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589147 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589168 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589242 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589208 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30589241 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589184 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589193 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589206 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589202 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589178 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589199 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589186 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589197 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589196 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589189 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589195 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589187 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589175 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589173 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589192 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589208 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12672583 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589203 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589190 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589204 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589185 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589188 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30589180 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10434526 - -
Affymetrix Mouse Gene 1.0 ST Array 10434524 - -
Affymetrix Mouse Gene 1.0 ST Array 10434527 - -
Affymetrix Mouse Gene 1.0 ST Array 10400392 - -
Affymetrix Mouse Gene 1.0 ST Array 10434525 - -
Affymetrix HT MG-430 PM Array Plate 1424473_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10434523 - -
Affymetrix Mouse Gene 1.1 ST Array 10400391 - -
Mouse Genome 430 2.0 Array 1424473_at - -
Mouse Genome 430 2.0 Array 1424473_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 245841_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 245841_at - -
Affymetrix Mouse Clariom S Array PSR1600002894.mm.1 NM_145632.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600002894.mm.1 NM_145632.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600002894.mm.1 TC1600000291.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600002893.mm.1 NM_145632.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600002893.mm.1 NM_145632.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600002893.mm.1 TC1600000291.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600002892.mm.1 NM_145632.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600002892.mm.1 NM_145632.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600002892.mm.1 TC1600000291.mm.1 MTA TC
Affymetrix HT Mouse Genome 430A Array 1424473_at - -
Affymetrix HT Mouse Genome 430A Array 1424473_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 5890242 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005890242 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772936 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2772936 - -
mouse-6 v1.1 (Illumina) scl0245841.4_4-S - -
Illumina Sentrix mouse-6.v1 bead array 5890242 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0245841.4_4-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890242 - -
MouseRef-8 v2 Expression BeadChip ILMN_2874443 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2874443 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772936 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0245841.4_4-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0