This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01344559_m1
See other N6AMT1 GE Assays ›
Gene Symbol:
N6amt1
Gene Name:
N-6 adenine-specific DNA methyltransferase 1 (putative)
Gene Aliases:
5830445C04Rik, Hemk2, Pred28
Chromosome Location:
Chr.16: 87354185 - 87368649 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01344559_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: N6amt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159331.1 NP_001152803.1 1 - 2 200 92
NM_026366.2 NP_080642.1 1 - 2 200 92
GenBank mRNA AK012019.1 - 1 - 2 140 92
AK145617.1 - 1 - 2 152 92
AK160298.1 - 1 - 2 162 92
AY456393.1 - 1 - 2 200 92
AY536887.1 - 1 - 2 132 92
AY583759.1 - 1 - 2 244 92
BC098330.1 - 1 - 2 142 30548526 92
BC116393.1 - 1 - 2 170 40101877 92
BC116394.1 - 1 - 2 170 40101881 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000952.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene N6amt1

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein methylation
positive regulation of cell growth
protein methyltransferase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600007416.mm.1 NM_001159331.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007416.mm.1 NM_001159331.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007416.mm.1 NM_026366.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007416.mm.1 NM_026366.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007416.mm.1 TC1600000952.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007421.mm.1 NM_001159331.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007421.mm.1 NM_001159331.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007421.mm.1 NM_026366.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007421.mm.1 NM_026366.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007421.mm.1 TC1600000952.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007420.mm.1 NM_001159331.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007420.mm.1 NM_001159331.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007420.mm.1 NM_026366.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007420.mm.1 NM_026366.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007420.mm.1 TC1600000952.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 909249 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0