This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01344563_m1
See other N6AMT1 GE Assays ›
Gene Symbol:
N6amt1
Gene Name:
N-6 adenine-specific DNA methyltransferase 1 (putative)
Gene Aliases:
5830445C04Rik, Hemk2, Pred28
Chromosome Location:
Chr.16: 87354185 - 87368649 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01344563_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: N6amt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026366.2 NP_080642.1 5 - 6 606 89
GenBank mRNA AK145617.1 - 5 - 6 558 89
AK160298.1 - 5 - 6 569 89
AY456393.1 - 5 - 6 606 89
BC116393.1 - 5 - 6 576 40101877 89
BC116394.1 - 5 - 6 576 40101881 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene N6amt1

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein methylation
positive regulation of cell growth
protein methyltransferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939274 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939276 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939275 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069192 AK012019.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043762 AY456393.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043762 AY583759.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043762 NM_001159331.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043762 NM_026366.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524110 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605050 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659111 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537539 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867303 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959723 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724201 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304880 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118857 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128578 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137284 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179541 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176125 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172578 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069192 AK012019.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043762 AY456393.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043762 AY583759.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043762 NM_001159331.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043762 NM_026366.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939292 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939286 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939257 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939276 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939251 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939278 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939266 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939277 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939269 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939289 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939250 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939280 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939254 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939283 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939290 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939275 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939294 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939263 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939293 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939261 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939271 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939268 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939273 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30939260 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179541 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172578 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176125 - -
Whole Mouse Genome G4122A A_51_P321033 BC116393.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P321033 BC116394.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P321033 AK160298.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P321033 AY456393.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P321033 AK145617.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123029 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172578 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176125 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179541 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12844906 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12844903 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12844905 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12959616 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939274 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939276 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30939275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939307 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939314 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939245 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939289 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939292 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939229 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939232 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939257 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939334 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939280 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939331 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30939251 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939289 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939264 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939292 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939249 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939276 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939299 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939232 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939266 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939280 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939278 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939286 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12684317 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939318 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939312 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12948995 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939293 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939261 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939316 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939313 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939236 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939309 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939307 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12659548 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939270 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939295 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939315 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939246 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12614064 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939269 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939281 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939297 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939243 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939257 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12614067 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939301 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939320 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939319 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939302 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939240 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939300 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939314 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939275 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12518298 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939260 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939298 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939263 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939228 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939250 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939237 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939283 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939273 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939235 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939271 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939254 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30939255 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10436703 - -
Affymetrix Mouse Gene 1.0 ST Array 10436701 - -
Affymetrix Mouse Gene 1.0 ST Array 10448092 - -
Affymetrix Mouse Gene 1.0 ST Array 10436706 - -
Affymetrix Mouse Gene 1.0 ST Array 10436707 - -
Affymetrix Mouse Gene 1.0 ST Array 10436700 - -
Affymetrix Mouse Gene 1.0 ST Array 10436702 - -
Affymetrix Mouse Gene 1.0 ST Array 10436699 - -
Affymetrix Mouse Gene 1.0 ST Array 10364911 - -
Affymetrix Mouse Gene 1.0 ST Array 10436705 - -
Affymetrix Mouse Gene 1.0 ST Array 10364910 - -
Affymetrix Mouse Gene 1.0 ST Array 10448093 - -
Affymetrix HT MG-430 PM Array Plate 1453821_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1430380_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10436698 - -
Mouse Genome 430 2.0 Array 1453821_PM_at - -
Mouse Genome 430 2.0 Array 1430380_PM_at - -
Mouse Genome 430 2.0 Array 1453821_at - -
Mouse Genome 430 2.0 Array 1430380_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67768_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67768_PM_at - -
Mouse Expression Set 430 1430380_at - -
Mouse Expression Set 430 1453821_at - -
Mouse Expression Set 430 1430380_PM_at - -
Mouse Expression Set 430 1453821_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430380_at - -
Affymetrix HT Mouse Genome 430B Array 1453821_at - -
Affymetrix HT Mouse Genome 430B Array 1430380_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453821_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 909249 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2340739 - -
Illumina mouse-6 v1.0 expression beadchip 2340739 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002340739 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2668289 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2668289 - -
mouse-6 v1.1 (Illumina) scl48912.5_69-S - -
Illumina Sentrix mouse-6.v1 bead array 2340739 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl48912.5_69-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340739 - -
MouseRef-8 v2 Expression BeadChip ILMN_2845391 - -
MouseRef-8 v2 Expression BeadChip ILMN_2668289 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2668289 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2845391 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl48912.5_69-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0