This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01344928_m1
See other PPIL2 GE Assays ›
Gene Symbol:
Ppil2
Gene Name:
peptidylprolyl isomerase (cyclophilin)-like 2
Gene Aliases:
0610009L05Rik, 1700016N17Rik, 4921520K19Rik, 4930511F14Rik, AA589416, C130078A06Rik, CYC4
Chromosome Location:
Chr.16: 17086555 - 17111263 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01344928_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppil2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252444.1 NP_001239373.1 1 - 2 105 92
NM_001252445.1 NP_001239374.1 1 - 2 105 92
NM_144954.3 NP_659203.1 1 - 2 105 92
GenBank mRNA AK009460.1 - 1 - 2 66 92
AK012472.1 - 1 - 2 91
AK081792.1 - 1 - 2 87 92
AK090052.1 - 1 - 2 98 92
AK161596.1 - 1 - 2 90 92
BY145175.1 - 1 - 2 102 92

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK009460.1, AK081792.1, AK090052.1, AK161596.1, NM_001252444.1, NM_001252445.1, NM_144954.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppil2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
protein peptidyl-prolyl isomerization
protein folding
protein ubiquitination
protein localization to plasma membrane
peptidyl-prolyl cis-trans isomerase activity
ubiquitin-protein transferase activity
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
isomerase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
ubiquitin-ubiquitin ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0