This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01345144_m1
See other USO1 GE Assays ›
Gene Symbol:
Uso1
Gene Name:
USO1 vesicle docking factor
Gene Aliases:
115kDa, TAP, Vdp
Chromosome Location:
Chr.5: 92137909 - 92202800 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01345144_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uso1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019490.1 NP_062363.1 3 - 4 380 158
GenBank mRNA AF096868.1 - 2 - 3 168 158
AK144638.1 - 3 - 4 371 158
AK156070.1 - 3 - 4 373 158
AK166587.1 - 4 - 5 340 158
AK172425.1 - 3 - 4 371 158
BC016069.1 - 3 - 4 380 4480992 158

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000868.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uso1

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
ER to Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
transcytosis
Golgi vesicle transport
Golgi vesicle docking
vesicle fusion with Golgi apparatus
membrane fusion
protein binding
protein transporter activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500007535.mm.1 TC0500000868.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500007535.mm.1 NM_019490.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500007535.mm.1 NM_019490.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0