This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01345700_m1
See other NDUFV3 GE Assays ›
Gene Symbol:
Ndufv3
Gene Name:
NADH dehydrogenase (ubiquinone) flavoprotein 3
Gene Aliases:
1500032D16Rik
Chromosome Location:
Chr.17: 31520115 - 31531326 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01345700_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufv3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083891.1 NP_001077360.1 1 - 2 151 107
NM_030087.2 NP_084363.2 1 - 2 151 107
GenBank mRNA AK018779.1 - 1 - 2 116 107
AK077433.1 - 1 - 2 92 107
AK154191.1 - 1 - 2 84 107
AK154477.1 - 1 - 2 130 107
AK160289.1 - 1 - 2 82 107
BC013640.1 - 1 - 2 108 4188234 107
BU555486.1 - 1 - 2 154 107
BY064612.1 - 1 - 2 119 107

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK018779.1, AK077433.1, AK154191.1, AK154477.1, AK160289.1, BC013640.1, NM_001083891.1, NM_030087.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufv3

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
oxidation-reduction process
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700004934.mm.1 NM_001083891.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004934.mm.1 NM_001083891.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004934.mm.1 NM_030087.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004934.mm.1 NM_030087.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700004934.mm.1 TC1700000539.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700004935.mm.1 NM_001083891.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004935.mm.1 NM_001083891.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004935.mm.1 NM_030087.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004935.mm.1 NM_030087.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700004935.mm.1 TC1700000539.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700004933.mm.1 NM_001083891.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004933.mm.1 NM_001083891.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004933.mm.1 NM_030087.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004933.mm.1 NM_030087.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700004933.mm.1 TC1700000539.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-svqt6:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0