This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01346428_m1
See other TRAT1 GE Assays ›
Gene Symbol:
Trat1
Gene Name:
T cell receptor associated transmembrane adaptor 1
Gene Aliases:
C030046M14Rik, Tcrim, Trim
Chromosome Location:
Chr.16: 48731914 - 48810520 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01346428_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198297.3 NP_938039.2 1 - 2 46 72
GenBank mRNA AJ297969.1 - 1 - 2 64 72
AK030807.1 - 1 - 2 229 72
AK138735.1 - 112 72
BC104350.1 - 1 - 2 46 40049557 72
BC104351.1 - 1 - 2 46 40049558 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001690.mm.1

Additional Information:

This assay is in the 5'UTR region of the AJ297969.1,BC104350.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trat1

Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of receptor recycling
immune system process
positive regulation of calcium-mediated signaling
positive regulation of T cell receptor signaling pathway
negative regulation of transport
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600013807.mm.1 NM_198297.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013807.mm.1 NM_198297.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013807.mm.1 TC1600001690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600013808.mm.1 NM_198297.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013808.mm.1 NM_198297.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013808.mm.1 TC1600001690.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 704051 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline