This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01346430_m1
See other TRAT1 GE Assays ›
Gene Symbol:
Trat1
Gene Name:
T cell receptor associated transmembrane adaptor 1
Gene Aliases:
C030046M14Rik, Tcrim, Trim
Chromosome Location:
Chr.16: 48731914 - 48810520 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01346430_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198297.3 NP_938039.2 3 - 4 194 140
GenBank mRNA AJ297969.1 - 3 - 4 212 140
AK030807.1 - 3 - 4 377 140
AK138735.1 - 260 140
BC104351.1 - 3 - 4 194 40049558 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001690.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trat1

Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of receptor recycling
immune system process
positive regulation of calcium-mediated signaling
positive regulation of T cell receptor signaling pathway
negative regulation of transport
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600013805.mm.1 NM_198297.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013805.mm.1 NM_198297.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013805.mm.1 TC1600001690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600013804.mm.1 NM_198297.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600013804.mm.1 NM_198297.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600013804.mm.1 TC1600001690.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline