This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01346922_m1
See other PTGR1 GE Assays ›
Gene Symbol:
Ptgr1
Gene Name:
prostaglandin reductase 1
Gene Aliases:
2510002C21Rik, Ltb4dh, Zadh3
Chromosome Location:
Chr.4: 58965590 - 58987131 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01346922_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptgr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025968.3 NP_080244.1 3 - 4 204 63
GenBank mRNA AK010888.1 - 3 - 4 239 63
AK011962.1 - 3 - 4 226 63
AK035425.1 - 3 - 4 205 63
AK134440.1 - 3 - 4 237 63
AK166835.1 - 3 - 4 204 63
BC014865.1 - 3 - 4 186 2648051 63
CT010267.1 - 2 - 3 153 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptgr1

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
zinc ion binding
oxidoreductase activity
enoyl-[acyl-carrier-protein] reductase activity
2,3-dihydroxy-2,3-dihydro-phenylpropionate dehydrogenase activity
cis-2,3-dihydrodiol DDT dehydrogenase activity
trans-9R,10R-dihydrodiolphenanthrene dehydrogenase activity
cis-chlorobenzene dihydrodiol dehydrogenase activity
2,5-dichloro-2,5-cyclohexadiene-1,4-diol dehydrogenase activity
trans-1,2-dihydrodiolphenanthrene dehydrogenase activity
2-alkenal reductase [NAD(P)] activity
3,4-dihydroxy-3,4-dihydrofluorene dehydrogenase activity
benzo(a)pyrene-trans-11,12-dihydrodiol dehydrogenase activity
benzo(a)pyrene-cis-4,5-dihydrodiol dehydrogenase activity
citronellyl-CoA dehydrogenase activity
menthone dehydrogenase activity
phthalate 3,4-cis-dihydrodiol dehydrogenase activity
13-prostaglandin reductase activity
cinnamate reductase activity
15-oxoprostaglandin 13-oxidase activity
NADPH-dependent curcumin reductase activity
NADPH-dependent dihydrocurcumin reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0