This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01347684_m1
See other SLC35A2 GE Assays ›
Gene Symbol:
Slc35a2
Gene Name:
solute carrier family 35 (UDP-galactose transporter), member A2
Gene Aliases:
AI327289, Had-1, Had1, Sfc8, UGT, Ugalt
Chromosome Location:
Chr.X: 7884034 - 7894492 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01347684_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc35a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_078484.2 NP_511039.2 4 - 5 1257 56
GenBank mRNA AF229634.1 - 4 - 5 1256 56
AK008095.1 - 3 - 4 389 56
CB723945.1 - 1 - 2 246 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000075.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc35a2

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
carbohydrate transport
nucleotide-sugar transport
UDP-galactose transmembrane transport
nucleotide-sugar transmembrane transporter activity
sugar:proton symporter activity
UDP-galactose transmembrane transporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00000585.mm.1 NM_001083937.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00000585.mm.1 NM_001083937.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00000585.mm.1 NM_078484.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00000585.mm.1 NM_078484.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00000585.mm.1 TC0X00000075.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00000594.mm.1 NM_078484.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00000594.mm.1 NM_078484.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00000594.mm.1 TC0X00000075.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00000591.mm.1 NM_078484.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00000591.mm.1 NM_078484.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00000591.mm.1 TC0X00000075.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 380079 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0