This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01348031_g1
See other FTSJ1 GE Assays ›
Gene Symbol:
Ftsj1
Gene Name:
FtsJ homolog 1 (E. coli)
Gene Aliases:
AI931847, Ftsj, Ftsjl, Sfc12
Chromosome Location:
Chr.X: 8238668 - 8252406 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01348031_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ftsj1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290430.1 NP_001277359.1 2 - 3 394 73
NM_133991.2 NP_598752.2 2 - 3 394 73
GenBank mRNA AK004357.1 - 2 - 3 352 73
AK036313.1 - 2 - 3 352 73
AK150251.1 - 2 - 3 196 73
BC034880.1 - 2 - 3 264 3468633 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001801.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ftsj1

Panther Classification:

Molecular Function -

RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00011594.mm.1 NM_133991.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011594.mm.1 NM_133991.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011594.mm.1 TC0X00001801.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011595.mm.1 NM_133991.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011595.mm.1 NM_133991.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011595.mm.1 TC0X00001801.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline