This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01348187_m1
See other PDE1C GE Assays ›
Gene Symbol:
Pde1c
Gene Name:
phosphodiesterase 1C
Gene Aliases:
-
Chromosome Location:
Chr.6: 56069798 - 56652516 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01348187_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025568.2 NP_001020739.1 6 - 7 1150 107
NM_001159952.1 NP_001153424.1 5 - 6 1123 107
NM_001159953.1 NP_001153425.1 6 - 7 1150 107
NM_001159955.1 NP_001153427.1 7 - 8 712 107
NM_001159956.1 NP_001153428.1 5 - 6 569 107
NM_001159957.1 NP_001153429.1 6 - 7 596 107
NM_001159960.1 NP_001153432.1 7 - 8 1024 107
NM_011054.4 NP_035184.1 7 - 8 1127 107
GenBank mRNA AK014887.1 - 5 - 6 540 107
AK015375.1 - 7 - 8 681 107
AK029531.1 - 6 - 7 594 107
AK030423.1 - 6 - 7 1152 107
AK050499.1 - 7 - 8 1025 107
AK077123.1 - 5 - 6 518 107
AK082180.1 - 6 - 7 1137 107
AK082497.1 - 6 - 7 935 107
AK133243.1 - 7 - 8 696 107
AK139923.1 - 5 - 6 1097 107
BB614593.1 - 5 - 6 570 107
BC032277.1 - 6 - 7 1079 5400689 107
L76944.1 - 6 - 7 780 107
L76946.1 - 7 - 8 1126 107
L76947.1 - 6 - 7 613 107

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde1c

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
sensory perception of smell
cGMP catabolic process
negative regulation of insulin secretion
response to calcium ion
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
calmodulin binding
phosphoric diester hydrolase activity
hydrolase activity
cAMP binding
metal ion binding
calcium- and calmodulin-regulated 3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline