This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01348254_m1
See other ABCG1 GE Assays ›
Gene Symbol:
Abcg1
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 1
Gene Aliases:
AW413978, Abc8, White
Chromosome Location:
Chr.17: 31057694 - 31117984 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01348254_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009593.2 NP_033723.1 10 - 11 1352 87
GenBank mRNA AF323659.1 - 10 - 11 1348 87
BC119471.2 - 10 - 11 1300 40048166 87
CV558133.1 - 2 - 3 163 87
U34920.1 - 10 - 11 1310 87
Z48745.1 - 10 - 11 1339 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000528.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcg1

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transport
lipid transport
cholesterol metabolic process
response to organic substance
negative regulation of macrophage derived foam cell differentiation
regulation of cholesterol esterification
positive regulation of cholesterol efflux
negative regulation of lipid storage
cholesterol transport
intracellular cholesterol transport
cholesterol efflux
phospholipid efflux
response to lipid
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
glycoprotein transport
cholesterol homeostasis
amyloid precursor protein catabolic process
reverse cholesterol transport
positive regulation of cholesterol biosynthetic process
transmembrane transport
phospholipid homeostasis
response to high density lipoprotein particle
toxin transport
nucleotide binding
ATP binding
phospholipid transporter activity
ATPase activity
cholesterol transporter activity
toxin transporter activity
sterol-transporting ATPase activity
glycoprotein transporter activity
protein homodimerization activity
ADP binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700004799.mm.1 NM_009593.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004799.mm.1 NM_009593.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700004799.mm.1 TC1700000528.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700004800.mm.1 NM_009593.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004800.mm.1 NM_009593.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700004800.mm.1 TC1700000528.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0