This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01348343_m1
See other SEC22C GE Assays ›
Gene Symbol:
Sec22c
Gene Name:
SEC22 vesicle trafficking protein homolog C (S. cerevisiae)
Gene Aliases:
4932412K21, 5930407I15Rik, C530046H07, Sec22l3
Chromosome Location:
Chr.9: 121680045 - 121706736 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01348343_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec22c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164562.1 NP_001158034.1 3 - 4 479 75
NM_178677.4 NP_848792.2 4 - 5 604 75
GenBank mRNA AK029985.1 - 4 - 5 535 75
AK031125.1 - 3 - 4 476 75
AK044249.1 - 3 - 4 479 75
BC046289.1 - 3 - 4 434 6469221 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003269.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec22c

Gene Ontology Categories:

Function(s) Process(es)

transport
ER to Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
vesicle fusion with Golgi apparatus
SNARE binding
SNAP receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900027564.mm.1 NM_001164562.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027564.mm.1 NM_001164562.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027564.mm.1 NM_178677.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027564.mm.1 NM_178677.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900027564.mm.1 TC0900003269.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cqsqq:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0