This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01348928_m1
See other EMC1 GE Assays ›
Gene Symbol:
Emc1
Gene Name:
ER membrane protein complex subunit 1
Gene Aliases:
2700016F22Rik
Chromosome Location:
Chr.4: 139352587 - 139378735 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01348928_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Emc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039200.2 NP_001034289.1 1 - 2 130 73
NM_146157.4 NP_666269.2 1 - 2 130 73
GenBank mRNA AK049061.1 - 1 - 2 121 73
AK077496.1 - 1 - 2 120 73
AK089289.1 - 1 - 2 109 73
AK129052.1 - 1 - 2 83 73
AK138637.1 - 1 - 2 102 73
AK161677.1 - 1 - 2 92 73
AK170739.1 - 1 - 2 109 73
BC129804.1 - 1 - 2 117 40087024 73
BC139366.1 - 1 - 2 116 8862388 73
BY281473.1 - 1 - 2 132 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004193.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Emc1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400014278.mm.1 NM_001039200.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014278.mm.1 NM_146157.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400014278.mm.1 TC0400004193.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400014279.mm.1 NM_001039200.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014279.mm.1 NM_146157.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400014279.mm.1 TC0400004193.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 849258 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lwb2x:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0