This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01348932_g1
See other EMC1 GE Assays ›
Gene Symbol:
Emc1
Gene Name:
ER membrane protein complex subunit 1
Gene Aliases:
2700016F22Rik
Chromosome Location:
Chr.4: 139352587 - 139378735 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01348932_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Emc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039200.2 NP_001034289.1 8 - 9 991 59
NM_146157.4 NP_666269.2 8 - 9 991 59
GenBank mRNA AK049061.1 - 8 - 9 982 59
AK077496.1 - 8 - 9 982 59
AK089289.1 - 8 - 9 970 59
AK129052.1 - 8 - 9 944 59
AK138637.1 - 8 - 9 963 59
AK161677.1 - 8 - 9 954 59
AK170739.1 - 8 - 9 970 59
BC048948.1 - 9 - 10 59
BC056952.1 - 3 - 4 439 59
BC129804.1 - 8 - 9 978 40087024 59
BC139366.1 - 8 - 9 977 8862388 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Emc1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0