This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01349421_m1
See other AUH GE Assays ›
Gene Symbol:
Auh
Gene Name:
AU RNA binding protein/enoyl-coenzyme A hydratase
Gene Aliases:
C77140, W91705
Chromosome Location:
Chr.13: 52835102 - 52929706 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01349421_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Auh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016709.2 NP_057918.2 8 - 9 853 70
GenBank mRNA AK003219.1 - 8 - 9 833 70
BC026525.1 - 8 - 9 791 70
BC049597.1 - 8 - 9 837 6593786 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Auh

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
metabolic process
branched-chain amino acid catabolic process
RNA binding
mRNA 3'-UTR binding
catalytic activity
enoyl-CoA hydratase activity
methylglutaconyl-CoA hydratase activity
lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0