This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01349467_m1
See other ECM2 GE Assays ›
Gene Symbol:
Ecm2
Gene Name:
extracellular matrix protein 2, female organ and adipocyte specific
Gene Aliases:
9030618O22Rik, BC065151, tenonectin
Chromosome Location:
Chr.13: 49504810 - 49532789 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01349467_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ecm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012324.2 NP_001012324.1 5 - 6 1304 139
GenBank mRNA AK038812.1 - 5 - 6 1166 139
AK040995.1 - 5 - 6 1328 139
AK134746.1 - 5 - 6 1258 139
AK163501.1 - 4 - 5 3326 139
BC065151.1 - 5 - 6 1174 5708030 139
BC089559.1 - 5 - 6 1304 30295705 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000565.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ecm2

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell-substrate adhesion
extracellular matrix organization
collagen binding
heparin binding
collagen V binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300003487.mm.1 NM_001012324.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300003487.mm.1 NM_001012324.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300003487.mm.1 TC1300000565.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-g928f:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0