This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01349524_g1
See other NSUN2 GE Assays ›
Gene Symbol:
Nsun2
Gene Name:
NOL1/NOP2/Sun domain family member 2
Gene Aliases:
D13Wsu123e, Misu
Chromosome Location:
Chr.13: 69612016 - 69635780 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01349524_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nsun2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145354.5 NP_663329.3 11 - 12 1300 81
GenBank mRNA AK030124.1 - 10 - 11 1185 81
AK075999.1 - 1 - 2 79 81
AK150631.1 - 11 - 12 1254 81
AK151917.1 - 11 - 12 1254 81
AK154789.1 - 11 - 12 1292 81
AK171399.1 - 11 - 12 1257 81
BC013625.1 - 7 - 8 772 4011674 81
BC023735.1 - 5 - 6 569 81
BC025549.1 - 11 - 12 1264 81
DQ490066.1 - 11 - 12 1263 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002766.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nsun2

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
tRNA processing
tRNA methylation
methylation
cell division
tRNA binding
RNA binding
methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
transferase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300005502.mm.1 NM_145354.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300005502.mm.1 TC1300002766.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0