Assay Details
| Gene Symbol: Ctsq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029636.3 | NP_083912.2 | 3 - 4 | 283 | 93 | |
| GenBank mRNA | AF456461.1 | - | 3 - 4 | 314 | 93 | |
| AY014776.1 | - | 3 - 4 | 283 | 93 | ||
| BC099415.1 | - | 3 - 4 | 290 | 30786803 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctsq
Genomic Map