This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01349992_m1
See other LSAMP GE Assays ›
Gene Symbol:
Lsamp
Gene Name:
limbic system-associated membrane protein
Gene Aliases:
5430428I19, AW046674, B130007O04Rik, D930023J12Rik, Lam, Lamp
Chromosome Location:
Chr.16: 41532788 - 42181679 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01349992_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lsamp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175548.3 NP_780757.1 2 - 3 428 76
GenBank mRNA AK030681.1 - 2 - 3 734 76
AK044845.1 - 2 - 3 428 76
AK140043.1 - 2 - 3 565 76
AK158702.1 - 2 - 3 912 76
AK164171.1 - 735 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000571.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lsamp

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s)

cell adhesion
single organismal cell-cell adhesion
locomotory exploration behavior
regulation of neurogenesis

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005388.mm.1 NM_175548.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005388.mm.1 NM_175548.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600005388.mm.1 TC1600000571.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline