This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01350505_m1
See other RBMS3 GE Assays ›
Gene Symbol:
Rbms3
Gene Name:
RNA binding motif, single stranded interacting protein
Gene Aliases:
6720477E09Rik, 8430436O14Rik
Chromosome Location:
Chr.9: 116572746 - 118040292 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01350505_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbms3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172122.1 NP_001165593.1 9 - 10 1561 53
NM_001172123.1 NP_001165594.1 9 - 10 1561 53
NM_001172124.1 NP_001165595.1 9 - 10 1561 53
NM_001172126.1 NP_001165597.1 9 - 10 1561 53
GenBank mRNA AK030609.1 - 9 - 10 1195 53
AK033005.1 - 9 - 10 1204 53
AK041098.1 - 9 - 10 1563 53
AK051776.1 - 10 - 11 1608 53
AK052196.1 - 9 - 10 1542 53

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003204.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of translation
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 NM_001172122.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 NM_001172122.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 NM_001172123.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 NM_001172123.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 NM_001172124.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 NM_001172124.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 NM_001172126.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 NM_001172126.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027024.mm.1 TC0900003204.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0