This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01350549_m1
See other LIPI GE Assays ›
Gene Symbol:
Lipi
Gene Name:
lipase, member I
Gene Aliases:
D930038D03Rik, lpd1
Chromosome Location:
Chr.16: 75540514 - 75586072 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01350549_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252513.1 NP_001239442.1 9 - 10 1278 111
GenBank mRNA AK079027.1 - 7 - 8 689 111
AK086576.1 - 9 - 10 1283 111
BC147403.1 - 4 - 5 470 9056446 111
BC147404.1 - 4 - 5 470 9056445 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001915.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
phospholipase activity
heparin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600014911.mm.1 NM_001252513.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014911.mm.1 NM_001252513.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014911.mm.1 TC1600001915.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0