This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01350786_m1
See other IGF2BP2 GE Assays ›
Gene Symbol:
Igf2bp2
Gene Name:
insulin-like growth factor 2 mRNA binding protein 2
Gene Aliases:
C330012H03Rik, IMP-2, Imp2, Neilsen
Chromosome Location:
Chr.16: 22059009 - 22163299 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01350786_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igf2bp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183029.2 NP_898850.2 13 - 14 1776 104
GenBank mRNA AK044984.1 - 12 - 13 1292 104
AK049196.1 - 13 - 14 1472 104
AK077404.1 - 13 - 14 1457 104
AK159654.1 - 13 - 14 1509 104
AK170531.1 - 12 - 13 1280 104
AY531659.1 - 13 - 14 1440 104
BC023758.1 - 13 - 14 1474 104
BU708344.1 - 3 - 4 221 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001375.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igf2bp2

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
transport
mRNA transport
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding
poly(A) RNA binding
mRNA 5'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600011380.mm.1 NM_183029.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011380.mm.1 NM_183029.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600011380.mm.1 TC1600001375.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wmcf5:80/100.66.77.18:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline