This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01351014_m1
See other TMPRSS3 GE Assays ›
Gene Symbol:
Tmprss3
Gene Name:
transmembrane protease, serine 3
Gene Aliases:
-
Chromosome Location:
Chr.17: 31179265 - 31209743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01351014_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163776.1 NP_001157248.1 3 - 4 400 131
NM_080727.2 NP_542765.2 2 - 3 260 131
GenBank mRNA AJ300738.1 - 2 - 3 260 131
AK158203.1 - 3 - 4 403 131
BC112375.1 - 2 - 3 257 40057032 131

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss3

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cellular sodium ion homeostasis
sensory perception of sound
serine-type endopeptidase activity
scavenger receptor activity
peptidase activity
serine-type peptidase activity
hydrolase activity
sodium channel regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-lmgjf:80/100.66.77.199:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline