This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01351015_g1
See other TMPRSS3 GE Assays ›
Gene Symbol:
Tmprss3
Gene Name:
transmembrane protease, serine 3
Gene Aliases:
-
Chromosome Location:
Chr.17: 31179265 - 31209743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01351015_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163776.1 NP_001157248.1 4 - 5 518 77
NM_080727.2 NP_542765.2 3 - 4 378 77
GenBank mRNA AK158203.1 - 4 - 5 521 77
BC112375.1 - 3 - 4 375 40057032 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001814.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss3

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cellular sodium ion homeostasis
sensory perception of sound
serine-type endopeptidase activity
scavenger receptor activity
peptidase activity
serine-type peptidase activity
hydrolase activity
sodium channel regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700016682.mm.1 NM_001163776.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016682.mm.1 NM_001163776.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016682.mm.1 NM_080727.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016682.mm.1 NM_080727.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700016682.mm.1 TC1700001814.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline