This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01351328_m1
See other N6AMT1 GE Assays ›
Gene Symbol:
N6amt1
Gene Name:
N-6 adenine-specific DNA methyltransferase 1 (putative)
Gene Aliases:
5830445C04Rik, Hemk2, Pred28
Chromosome Location:
Chr.16: 87354185 - 87368649 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01351328_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: N6amt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159331.1 NP_001152803.1 4 - 5 467 96
GenBank mRNA AY583759.1 - 4 - 5 511 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000952.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein methylation
positive regulation of cell growth
protein methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600007428.mm.1 NM_001159331.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007428.mm.1 NM_001159331.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007428.mm.1 NM_026366.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007428.mm.1 NM_026366.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007428.mm.1 TC1600000952.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007430.mm.1 NM_001159331.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007430.mm.1 NM_001159331.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007430.mm.1 NM_026366.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007430.mm.1 NM_026366.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007430.mm.1 TC1600000952.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007431.mm.1 NM_001159331.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007431.mm.1 NM_001159331.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007431.mm.1 NM_026366.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007431.mm.1 NM_026366.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007431.mm.1 TC1600000952.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0