This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01351742_g1
See other MYD88 GE Assays ›
Gene Symbol:
Myd88
Gene Name:
myeloid differentiation primary response gene 88
Gene Aliases:
-
Chromosome Location:
Chr.9: 119335988 - 119340040 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01351742_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Myd88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010851.2 NP_034981.1 3 - 4 731 88
GenBank mRNA AK150083.1 - 3 - 4 614 88
AK151748.1 - 3 - 4 687 88
AK152596.1 - 3 - 4 716 88
AK152935.1 - 3 - 4 689 88
AK167929.1 - 3 - 4 711 88
BC005591.1 - 3 - 4 731 3482246 88
BC051075.1 - 3 - 4 678 88
BC058787.1 - 3 - 4 666 6309465 88
DQ790885.1 - 3 - 4 650 88
DQ790886.1 - 3 - 4 650 88
U84409.1 - 3 - 4 712 88
X51397.1 - 3 - 4 600 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003241.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Myd88

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

response to molecule of fungal origin
immune system process
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
immune response
signal transduction
cell surface receptor signaling pathway
transmembrane receptor protein serine/threonine kinase signaling pathway
JNK cascade
Toll signaling pathway
response to virus
immunoglobulin mediated immune response
cytokine-mediated signaling pathway
lipopolysaccharide-mediated signaling pathway
response to peptidoglycan
response to lipopolysaccharide
positive regulation of interleukin-17 production
positive regulation of interleukin-23 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
regulation of cell proliferation
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of growth of symbiont in host
positive regulation of chemokine biosynthetic process
innate immune response
type I interferon biosynthetic process
positive regulation of JNK cascade
positive regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
regulation of inflammatory response
defense response to Gram-positive bacterium
positive regulation of NF-kappaB transcription factor activity
response to interleukin-1
3'-UTR-mediated mRNA stabilization
establishment of endothelial intestinal barrier
receptor binding
Toll binding
interleukin-1 receptor binding
protein binding
identical protein binding
TIR domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900027274.mm.1 NM_010851.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027274.mm.1 NM_010851.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900027274.mm.1 TC0900003241.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0