This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01351833_m1
See other PHACTR1 GE Assays ›
Gene Symbol:
Phactr1
Gene Name:
phosphatase and actin regulator 1
Gene Aliases:
9630030F18Rik, Rpel1
Chromosome Location:
Chr.13: 42680571 - 43138526 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01351833_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phactr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005740.1 NP_001005740.1 2 - 3 433 83
GenBank mRNA AK048208.1 - 2 - 3 329 83
AK139452.1 - 2 - 3 405 83
AY993932.1 - 4 - 5 577 83
BC048407.1 - 2 - 3 471 83
BC061691.1 - 2 - 3 433 6307669 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000482.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phactr1

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

actomyosin structure organization
actin cytoskeleton reorganization
regulation of phosphorylation
stress fiber assembly
cell motility
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300002900.mm.1 NM_001005740.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002900.mm.1 NM_001005740.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002900.mm.1 TC1300000482.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline