Assay Details
| Gene Symbol: Upf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122829.1 | NP_001116301.1 | 6 - 7 | 1241 | 107 | |
| NM_030680.2 | NP_109605.2 | 6 - 7 | 1241 | 107 | ||
| GenBank mRNA | AF182947.1 | - | 7 - 8 | 1230 | 107 | |
| AF322655.1 | - | 6 - 7 | 961 | 107 | ||
| AK148196.1 | - | 7 - 8 | 1218 | 107 | ||
| BC052149.1 | - | 6 - 7 | 1179 | 6332288 | 107 | |
| BC056442.1 | - | 6 - 7 | 1241 | 5702493 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upf1
Genomic Map