This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01352798_m1
See other SMC2 GE Assays ›
Gene Symbol:
Smc2
Gene Name:
structural maintenance of chromosomes 2
Gene Aliases:
5730502P04Rik, AI255214, AW545314, CAP-E, CAPE, Fin16, SMC-2, Smc2l1
Chromosome Location:
Chr.4: 52439221 - 52488365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01352798_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301412.1 NP_001288341.1 16 - 17 2381 89
NM_008017.4 NP_032043.3 16 - 17 2305 89
GenBank mRNA AJ534939.1 - 15 - 16 2136 89
AK028233.1 - 16 - 17 2348 89
AK145341.1 - 16 - 17 2335 89
AK152090.1 - 16 - 17 2333 89
BC094380.1 - 16 - 17 2277 30543190 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smc2

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
mitotic chromosome condensation
meiotic chromosome condensation
chromosome condensation
meiotic chromosome segregation
chromosome organization
cell division
kinetochore organization
nucleotide binding
protein binding
ATP binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gcsl6:80/100.66.76.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0