This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01353556_m1
See other PALLD GE Assays ›
Gene Symbol:
Palld
Gene Name:
palladin, cytoskeletal associated protein
Gene Aliases:
2410003B16Rik, 6030492A02
Chromosome Location:
Chr.8: 61511429 - 61902700 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01353556_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Palld
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081390.1 NP_001074859.1 3 - 4 1333 66
NM_001293772.1 NP_001280701.1 3 - 4 1337 66
NM_001293773.1 NP_001280702.1 2 - 3 468 66
GenBank mRNA AK031696.1 - 2 - 3 460 66
AK052489.1 - 3 - 4 1315 66
AK076423.1 - 1 - 2 105 66
AK087270.1 - 2 - 3 470 66
AK162050.1 - 2 - 3 460 66
AK173081.1 - 2 - 3 408 66
JX477684.1 - 2 - 3 460 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the JX477684.1, NM_001293773.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

keratinocyte development
epithelial cell morphogenesis
actin cytoskeleton organization
neuron projection development
ruffle organization
positive regulation of podosome assembly
actin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0