This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01354262_m1
See other SRD5A1 GE Assays ›
Gene Symbol:
Srd5a1
Gene Name:
steroid 5 alpha-reductase 1
Gene Aliases:
0610031P22Rik, 4930435F02Rik, S5AR 1, Srd5a-1
Chromosome Location:
Chr.13: 69573449 - 69611900 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01354262_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srd5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC094503.1 - 645 5053446 80
BC115468.1 - 2 - 3 523 40100423 80
BC115469.1 - 2 - 3 40100424 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002276.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300014872.mm.1 TC1300002276.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014871.mm.1 TC1300002276.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014874.mm.1 NM_175283.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014874.mm.1 NM_175283.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300014874.mm.1 TC1300002276.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014873.mm.1 TC1300002276.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0