This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01354263_m1
See other SRD5A1 GE Assays ›
Gene Symbol:
Srd5a1
Gene Name:
steroid 5 alpha-reductase 1
Gene Aliases:
0610031P22Rik, 4930435F02Rik, S5AR 1, Srd5a-1
Chromosome Location:
Chr.13: 69573449 - 69611900 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01354263_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srd5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175283.3 NP_780492.2 3 - 4 764 100
GenBank mRNA BC079863.1 - 3 - 4 742 6807475 100
BC094503.1 - 743 5053446 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002276.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Srd5a1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
steroid biosynthetic process
androgen biosynthetic process
sex differentiation
steroid metabolic process
androgen metabolic process
cell differentiation
progesterone metabolic process
response to drug
oxidation-reduction process
3-oxo-5-alpha-steroid 4-dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
amide binding
cholestenone 5-alpha-reductase activity
NADPH binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300014869.mm.1 NM_175283.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014869.mm.1 NM_175283.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300014869.mm.1 TC1300002276.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-b5lz9:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0