This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01354525_m1
See other TRMT2B GE Assays ›
Gene Symbol:
Trmt2b
Gene Name:
TRM2 tRNA methyltransferase 2B
Gene Aliases:
4732479N06Rik
Chromosome Location:
Chr.X: 134222955 - 134277039 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01354525_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trmt2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167994.1 NP_001161466.1 6 - 7 729 98
NM_172540.2 NP_766128.2 5 - 6 653 98
GenBank mRNA AK028999.1 - 5 - 6 609 98
AK049523.1 - 5 - 6 651 98
AK129030.1 - 6 - 7 672 98
AK137595.1 - 615 98
BC030387.1 - 4 - 5 314 98
BC056937.1 - 6 - 7 670 5716144 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002990.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trmt2b

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
tRNA processing
biological_process
methylation
methyltransferase activity
RNA methyltransferase activity
transferase activity
S-adenosylmethionine-dependent tRNA (m5U54) methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00018620.mm.1 NM_001167994.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018620.mm.1 NM_001167994.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018620.mm.1 NM_172540.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018620.mm.1 NM_172540.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00018620.mm.1 TC0X00002990.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0