This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01402432_m1
See other GDNF GE Assays ›
Gene Symbol:
Gdnf
Gene Name:
glial cell line derived neurotrophic factor
Gene Aliases:
AI385739
Chromosome Location:
Chr.15: 7810048 - 7837580 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01402432_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gdnf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301332.1 NP_001288261.1 2 - 3 1165 91
GenBank mRNA U75532.1 - 2 - 3 282 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

neurotrophic factor

Gene Ontology Categories:

Function(s) Process(es)

metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
neural crest cell migration
organ induction
postsynaptic membrane organization
mesenchymal to epithelial transition involved in metanephros morphogenesis
transmembrane receptor protein tyrosine kinase signaling pathway
transforming growth factor beta receptor signaling pathway
nervous system development
peripheral nervous system development
cell proliferation
positive regulation of cell proliferation
response to wounding
regulation of gene expression
postganglionic parasympathetic fiber development
neuron differentiation
peristalsis
neuron projection development
positive regulation of monooxygenase activity
negative regulation of neuron apoptotic process
positive regulation of cell differentiation
positive regulation of transcription from RNA polymerase II promoter
mRNA stabilization
enteric nervous system development
sympathetic nervous system development
regulation of dopamine uptake involved in synaptic transmission
ureteric bud formation
regulation of morphogenesis of a branching structure
cellular response to dexamethasone stimulus
regulation of ureteric bud formation
positive regulation of ureteric bud formation
positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
transforming growth factor beta receptor binding
growth factor activity
glial cell-derived neurotrophic factor receptor binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8mgvv:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0