This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01490249_m1
See other TMEM9 GE Assays ›
Gene Symbol:
Tmem9
Gene Name:
transmembrane protein 9
Gene Aliases:
1500015G18Rik, AW545782
Chromosome Location:
Chr.1: 136008219 - 136035030 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01490249_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160145.1 NP_001153617.1 3 - 4 389 64
NM_001160146.1 NP_001153618.1 3 - 4 285 64
NM_025439.3 NP_079715.1 3 - 4 254 64
GenBank mRNA AK005248.1 - 3 - 4 241 64
AK021225.1 - 3 - 4 241 64
AK134909.1 - 3 - 4 219 64
AK154423.1 - 3 - 4 241 64
AK218177.1 - 1 - 2 56 64
BC016524.1 - 3 - 4 229 3594475 64
BC020291.1 - 2 - 3 206 64
BC048017.1 - 3 - 4 208 64
BQ918002.1 - 3 - 4 476 64
CA460035.1 - 3 - 4 291 64
CX208138.1 - 3 - 4 285 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem9

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline