This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01545271_g1
See other GTF2F1 GE Assays ›
Gene Symbol:
Gtf2f1
Gene Name:
general transcription factor IIF, polypeptide 1
Gene Aliases:
2810405L04Rik, C76800, RAP74
Chromosome Location:
Chr.17: 57003401 - 57011393 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01545271_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2f1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133801.2 NP_598562.1 6 - 7 853 77
GenBank mRNA AK013002.1 - 6 - 7 867 77
AK168236.1 - 6 - 7 866 77
AK168243.1 - 6 - 7 866 77
BC003336.1 - 6 - 7 850 77
BC023081.1 - 6 - 7 853 5355475 77
BC031123.1 - 6 - 7 983 5026448 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2f1

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
positive regulation of transcription elongation from RNA polymerase II promoter
positive regulation of catalytic activity
positive regulation of transcription from RNA polymerase II promoter
DNA binding
catalytic activity
protein binding
transcription factor binding
phosphatase activator activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0