This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01546135_m1
See other TRPS1 GE Assays ›
Gene Symbol:
Trps1
Gene Name:
trichorhinophalangeal syndrome I (human)
Gene Aliases:
AI115454, AI447310, D15Ertd586e
Chromosome Location:
Chr.15: 50654757 - 50890747 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01546135_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trps1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC037058.1 - 1 - 2 118 4951294 117

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC037058.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500011363.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011363.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011363.mm.1 TC1500001491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500011370.mm.1 TC1500001491.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 717706 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0