This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01598982_g1
See other TWF1 GE Assays ›
Gene Symbol:
Twf1
Gene Name:
twinfilin, actin-binding protein, homolog 1 (Drosophila)
Gene Aliases:
A6, Ptk9, twinfilin
Chromosome Location:
Chr.15: 94577948 - 94589884 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01598982_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Twf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008971.4 NP_032997.3 4 - 5 426 89
GenBank mRNA AK077410.1 - 4 - 5 482 89
AK147990.1 - 4 - 5 420 89
AK150852.1 - 4 - 5 449 89
AK165561.1 - 3 - 4 356 89
AK168142.1 - 4 - 5 444 89
AY267188.1 - 4 - 5 378 89
BC015081.1 - 4 - 5 443 3979221 89
BC094034.1 - 4 - 5 426 4934593 89
U82324.1 - 4 - 5 434 89

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Twf1

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-tyrosine phosphorylation
negative regulation of actin filament polymerization
sequestering of actin monomers
regulation of actin phosphorylation
barbed-end actin filament capping
actin binding
actin monomer binding
protein tyrosine kinase activity
ATP binding
phosphatidylinositol-4,5-bisphosphate binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016987.mm.1 NM_008971.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016987.mm.1 NM_008971.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500016987.mm.1 TC1500002111.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016988.mm.1 NM_008971.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016988.mm.1 NM_008971.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500016988.mm.1 TC1500002111.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016986.mm.1 NM_008971.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016986.mm.1 NM_008971.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500016986.mm.1 TC1500002111.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0