| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100018477.mm.1 |
NM_028865.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018477.mm.1 |
NM_028865.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018477.mm.1 |
TC1100004281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018473.mm.1 |
NM_028865.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018473.mm.1 |
NM_028865.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018473.mm.1 |
TC1100004281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018479.mm.1 |
NM_028865.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018479.mm.1 |
NM_028865.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018479.mm.1 |
TC1100004281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018484.mm.1 |
NM_028865.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018484.mm.1 |
NM_028865.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018484.mm.1 |
TC1100004281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100018486.mm.1 |
NM_028865.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018486.mm.1 |
NM_028865.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100018486.mm.1 |
TC1100004281.mm.1 |
MTA TC |