This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01606499_g1
See other ROMO1 GE Assays ›
Gene Symbol:
Romo1
Gene Name:
reactive oxygen species modulator 1
Gene Aliases:
2010100O12Rik, AI853864
Chromosome Location:
Chr.2: 156144153 - 156145794 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01606499_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Romo1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164216.1 NP_001157688.1 1 - 2 125 110
GenBank mRNA AA058111.1 - 1 - 2 125 110
AK008300.1 - 1 - 2 60 110

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK008300.1, NM_001164216.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

replicative cell aging
positive regulation of cell proliferation
cellular response to reactive oxygen species
defense response to bacterium
positive regulation of reactive oxygen species metabolic process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200019157.mm.1 NM_001164216.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200019157.mm.1 NM_001164216.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200019157.mm.1 NM_001164217.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200019157.mm.1 NM_025946.6 Refseq
Affymetrix Mouse Clariom S Array PSR0200019157.mm.1 TC0200002384.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0