This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01610405_g1
See other UXT GE Assays ›
Gene Symbol:
Uxt
Gene Name:
ubiquitously expressed transcript
Gene Aliases:
0910002B17Rik
Chromosome Location:
Chr.X: 20951665 - 20962035 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01610405_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uxt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK149612.1 - 1258 152
AK149753.1 - 1258 152
AK149952.1 - 1258 152
AK150172.1 - 1258 152
AK163983.1 - 1 - 2 87 152

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK163983.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 716195 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0