This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01610408_gH
See other UXT GE Assays ›
Gene Symbol:
Uxt
Gene Name:
ubiquitously expressed transcript
Gene Aliases:
0910002B17Rik
Chromosome Location:
Chr.X: 20951665 - 20962035 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01610408_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uxt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK086822.1 - 4 - 5 314 160
BC029258.1 - 4 - 5 288 4483276 160

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00003420.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00012297.mm.1 NM_013840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012297.mm.1 NM_013840.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012297.mm.1 TC0X00003420.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00012299.mm.1 NM_013840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012299.mm.1 NM_013840.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012299.mm.1 TC0X00003420.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00012298.mm.1 NM_013840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012298.mm.1 NM_013840.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012298.mm.1 TC0X00003420.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00012302.mm.1 NM_013840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012302.mm.1 NM_013840.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012302.mm.1 TC0X00003420.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00012296.mm.1 TC0X00003420.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 624742 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0