This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01611012_m1
See other CEACAM1 GE Assays ›
Gene Symbol:
Ceacam1
Gene Name:
carcinoembryonic antigen-related cell adhesion molecule 1
Gene Aliases:
Bgp, Bgp1, C-CAM, CD66a, Cc1, Cea-1, Cea-7, Cea1, Cea7, Hv-2, Hv2, MHVR, MHVR1, Mhv-1, bb-1, mCEA1, mmCGM1, mmCGM1a, mmCGM2
Chromosome Location:
Chr.7: 25461702 - 25477625 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01611012_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ceacam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039187.1 NP_001034276.1 2 - 3 518 121
NM_011926.2 NP_036056.2 2 - 3 518 121
GenBank mRNA AB236328.2 - 2 - 3 427 121
AB236329.1 - 2 - 3 427 121
AB236331.2 - 2 - 3 427 121
AK085408.1 - 641 121
X67278.1 - 2 - 3 530 121
X67282.1 - 2 - 3 530 121

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ceacam1

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cytokine production
homophilic cell adhesion via plasma membrane adhesion molecules
viral process
calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
negative regulation of interleukin-2 production
positive regulation of T cell proliferation
negative regulation of T cell proliferation
positive regulation of MAP kinase activity
negative regulation by host of viral process
positive regulation by host of viral process
negative regulation by host of viral genome replication
negative regulation of JNK cascade
positive regulation of JNK cascade
viral entry into host cell
Peyer's patch development
positive regulation of immunoglobulin secretion
positive regulation of NFAT protein import into nucleus
positive regulation of activation-induced cell death of T cells
positive regulation of CD4-positive, alpha-beta T cell activation
positive regulation of CD4-positive, alpha-beta T cell proliferation
positive regulation of CD8-positive, alpha-beta T cell activation
virus receptor activity
protein binding
protein kinase binding
protein homodimerization activity
virion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline